Product Details

SNP ID
rs192028283
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:48032173 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCTCACCCGCCATCCACAATCTC[C/G]TCCAGCACCAAGAAGGCTCCGTCCA
Phenotype
MIM: 615526 MIM: 613788
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
COPZ2 PubMed Links

Gene Details

Gene
COPZ2
Gene Name
coatomer protein complex subunit zeta 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016429.3 506 Silent Mutation GAC,GAG D159E NP_057513.1
XM_011524874.2 506 Intron XP_011523176.1
Gene
MIR152
Gene Name
microRNA 152
There are no transcripts associated with this gene.

View Full Product Details