Product Details

SNP ID
rs191981941
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:8522457 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGGCCCACGTACTGGTATAGGGCC[C/T]GGCACCTGGGACCATGTGTCCGAGG
Phenotype
MIM: 601480
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MYO1F PubMed Links

Gene Details

Gene
MYO1F
Gene Name
myosin IF
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012335.3 3206 Missense Mutation CAG,CGG Q1047R NP_036467.2
XM_011528024.1 3206 Missense Mutation CAG,CGG Q1068R XP_011526326.1
XM_011528025.1 3206 Missense Mutation CAG,CGG Q1064R XP_011526327.1
XM_011528026.1 3206 Missense Mutation CAG,CGG Q1039R XP_011526328.1
XM_011528027.1 3206 Missense Mutation CAG,CGG Q1023R XP_011526329.1
XM_011528028.2 3206 Intron XP_011526330.1
XM_017026821.1 3206 Missense Mutation CAG,CGG Q1043R XP_016882310.1
Gene
ZNF414
Gene Name
zinc finger protein 414
There are no transcripts associated with this gene.

View Full Product Details