Product Details

SNP ID
rs191813214
Assay Type
Functionally tested
NCBI dbSNP Submissions
7
Location
Chr.1:37568974 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCACATTTTCCTCCTCAGGCTCC[A/G]AGGAAGACTCTTCCGCATCATCTCT
Phenotype
MIM: 602135 MIM: 609365
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DNALI1 PubMed Links

Gene Details

Gene
DNALI1
Gene Name
dynein axonemal light intermediate chain 1
There are no transcripts associated with this gene.

Gene
GNL2
Gene Name
G protein nucleolar 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001323623.1 1908 Missense Mutation TCG,TTG S650L NP_001310552.1
NM_001323624.1 1908 Missense Mutation TCG,TTG S399L NP_001310553.1
NM_013285.2 1908 Missense Mutation TCG,TTG S582L NP_037417.1

View Full Product Details