Product Details

SNP ID
rs191761952
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:112647441 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGGCAAAGGGGCAGGAGGGTGTCA[A/G]GTGGTCTGAACTGATAAAAATTGGT
Phenotype
MIM: 137570
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FLJ42351 PubMed Links

Gene Details

Gene
FLJ42351
Gene Name
uncharacterized LOC400999
There are no transcripts associated with this gene.

Gene
SLC20A1
Gene Name
solute carrier family 20 member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005415.4 962 Missense Mutation AAG,AGG K151R NP_005406.3
XM_017004768.1 962 UTR 5 XP_016860257.1
XM_017004769.1 962 UTR 5 XP_016860258.1
XM_017004770.1 962 UTR 5 XP_016860259.1
XM_017004771.1 962 UTR 5 XP_016860260.1

View Full Product Details