Product Details

SNP ID
rs191813843
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:36749610 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGATTTTCCACATTCCTTACAACAA[C/T]AGGGTTTCTCACCAGTGTGAATCCG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF850 PubMed Links

Gene Details

Gene
ZNF850
Gene Name
zinc finger protein 850
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193552.1 1493 Missense Mutation TAT,TGT Y477C NP_001180481.1
NM_001267779.1 1493 Missense Mutation TAT,TGT Y445C NP_001254708.1

View Full Product Details