Product Details

SNP ID
rs191816066
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:120112901 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGTCTTCAGTCACACCTAAGTTTT[C/T]GGCAAGTTCCCTTCTGTGGAGAGAA
Phenotype
MIM: 300446
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LINC01402 PubMed Links

Gene Details

Gene
LINC01402
Gene Name
long intergenic non-protein coding RNA 1402
There are no transcripts associated with this gene.

Gene
RHOXF1
Gene Name
Rhox homeobox family member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_139282.2 538 Missense Mutation AAA,GAA K138E NP_644811.1
XM_011531281.2 538 Missense Mutation AAA,GAA K166E XP_011529583.1
Gene
RHOXF1-AS1
Gene Name
RHOXF1 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details