Product Details

SNP ID
rs191765580
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:59457016 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGTTTTTCTTCCTGGAACTTACAC[A/G]TTCCCGAGAGCTGGAGTTTTTCTTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR4D6 PubMed Links

Gene Details

Gene
OR4D6
Gene Name
olfactory receptor family 4 subfamily D member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004708.1 56 Missense Mutation CAT,CGT H19R NP_001004708.1

View Full Product Details