Product Details

SNP ID
rs191879618
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:104457467 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGAACAGAATCTGCCGGATGGTGC[C/T]GGCCACTTGCTTGGGATCCTTTATC
Phenotype
MIM: 610128
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CHST11 PubMed Links

Gene Details

Gene
CHST11
Gene Name
carbohydrate (chondroitin 4) sulfotransferase 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001173982.1 554 Missense Mutation CCG,CTG P19L NP_001167453.1
NM_018413.5 554 Missense Mutation CCG,CTG P19L NP_060883.1
XM_017019369.1 554 Intron XP_016874858.1

View Full Product Details