Product Details

SNP ID
rs191891365
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:27598106 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTCCATAAAGGATAATAGTTTCCA[A/G]TACACTATCCCTCATGATGACTCCT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF512 PubMed Links

Gene Details

Gene
ZNF512
Gene Name
zinc finger protein 512
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271286.1 270 Silent Mutation CAA,CAG Q42Q NP_001258215.1
NM_001271287.1 270 UTR 5 NP_001258216.1
NM_001271288.1 270 UTR 5 NP_001258217.1
NM_001271289.1 270 UTR 5 NP_001258218.1
NM_001271318.1 270 UTR 5 NP_001258247.1
NM_032434.3 270 Silent Mutation CAA,CAG Q43Q NP_115810.2

View Full Product Details