Product Details

SNP ID
rs191891370
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:143726500 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTGGCCGTTCTCCTGCGGCACTGG[A/G]AAGCCACCCTCATCCTCCATGCGCC
Phenotype
MIM: 611927
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM83H PubMed Links

Gene Details

Gene
FAM83H
Gene Name
family with sequence similarity 83 member H
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198488.3 4226 Silent Mutation TTC,TTT F987F NP_940890.3
XM_005250887.3 4226 Silent Mutation TTC,TTT F1006F XP_005250944.1
XM_005250888.3 4226 Silent Mutation TTC,TTT F993F XP_005250945.1
XM_005250889.3 4226 Silent Mutation TTC,TTT F987F XP_005250946.1
XM_011516980.2 4226 Silent Mutation TTC,TTT F1188F XP_011515282.2
XM_011516981.2 4226 Silent Mutation TTC,TTT F1043F XP_011515283.1
Gene
FAM83H-AS1
Gene Name
FAM83H antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

Gene
MAPK15
Gene Name
mitogen-activated protein kinase 15
There are no transcripts associated with this gene.

Gene
MIR4664
Gene Name
microRNA 4664
There are no transcripts associated with this gene.

View Full Product Details