Product Details

SNP ID
rs191843509
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:94208412 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTACCAATATGAATCTTTTCAACCT[G/T]GACCGTTTTCGCTTTGAGAAAAGGA
Phenotype
MIM: 612761
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LOC101929210 PubMed Links

Gene Details

Gene
LOC101929210
Gene Name
uncharacterized LOC101929210
There are no transcripts associated with this gene.

Gene
SMARCAD1
Gene Name
SWI/SNF-related, matrix-associated actin-dependent regulator of chromatin, subfamily a, containing DEAD/H box 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128429.2 117 Silent Mutation CTG,CTT L6L NP_001121901.1
NM_001128430.1 117 Silent Mutation CTG,CTT L6L NP_001121902.1
NM_001254949.1 117 Intron NP_001241878.1
NM_020159.4 117 Silent Mutation CTG,CTT L6L NP_064544.2
XM_017008463.1 117 Silent Mutation CTG,CTT L6L XP_016863952.1
XM_017008464.1 117 UTR 5 XP_016863953.1
XM_017008465.1 117 UTR 5 XP_016863954.1

View Full Product Details