Product Details

SNP ID
rs191920635
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:233840650 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGTGAAGAGCTGCCCTGGTCTCC[C/T]GGTCTGACGGTGTTGTCCACCCCAT
Phenotype
MIM: 612667
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HJURP PubMed Links

Gene Details

Gene
HJURP
Gene Name
Holliday junction recognition protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282962.1 2196 Silent Mutation CCA,CCG P656P NP_001269891.1
NM_001282963.1 2196 Silent Mutation CCA,CCG P625P NP_001269892.1
NM_018410.4 2196 Silent Mutation CCA,CCG P710P NP_060880.3
XM_011511437.1 2196 Silent Mutation CCA,CCG P598P XP_011509739.1
Gene
MROH2A
Gene Name
maestro heat like repeat family member 2A
There are no transcripts associated with this gene.

View Full Product Details