Product Details

SNP ID
rs192263732
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:30941030 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGCCGGTCCACAGCCTCTGCTCC[A/G]CCTTTGGCCTCCAGTGCTGCCTCCT
Phenotype
MIM: 613928
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DPCR1 PubMed Links

Gene Details

Gene
DPCR1
Gene Name
diffuse panbronchiolitis critical region 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080870.3 31 Missense Mutation ACC,GCC T11A NP_543146.2
Gene
SFTA2
Gene Name
surfactant associated 2
There are no transcripts associated with this gene.

View Full Product Details