Product Details

SNP ID
rs192257012
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:122534686 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGCCTCATTAAATCCAAAGATAC[A/G]GGCATGCAGCTTAAGTGATGGGTTT
Phenotype
MIM: 607363
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KNTC1 PubMed Links

Gene Details

Gene
KNTC1
Gene Name
kinetochore associated 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014708.4 297 Missense Mutation CAG,CGG Q51R NP_055523.1
XM_006719706.2 297 Missense Mutation CAG,CGG Q51R XP_006719769.1
XM_011539027.2 297 Missense Mutation CAG,CGG Q51R XP_011537329.1
XM_011539029.2 297 Missense Mutation CAG,CGG Q51R XP_011537331.1
XM_011539030.1 297 Missense Mutation CAG,CGG Q51R XP_011537332.1
XM_017020254.1 297 Missense Mutation CAG,CGG Q51R XP_016875743.1
XM_017020255.1 297 UTR 5 XP_016875744.1
XM_017020256.1 297 Intron XP_016875745.1
XM_017020257.1 297 Missense Mutation CAG,CGG Q51R XP_016875746.1
Gene
RSRC2
Gene Name
arginine and serine rich coiled-coil 2
There are no transcripts associated with this gene.

View Full Product Details