Product Details

SNP ID
rs192278505
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:77590076 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGATGCTGTCGGTGGTGCTGCTCAT[A/G]GGGCTGGCCCGCGTAGTCGCCCGGC
Phenotype
MIM: 609914
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AQP11 PubMed Links

Gene Details

Gene
AQP11
Gene Name
aquaporin 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173039.2 423 Missense Mutation ATA,ATG I28M NP_766627.1
XM_005273917.4 423 Missense Mutation ATA,ATG I28M XP_005273974.1
Gene
LOC107987163
Gene Name
uncharacterized LOC107987163
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_017018644.1 423 Intron XP_016874133.1

View Full Product Details