Product Details

SNP ID
rs192282834
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:12664979 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTATCGGTGAACAATTTTCTCGGC[A/G]GTGACCACCACGACTCGGAGCCCGT
Phenotype
MIM: 615603
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CPPED1 PubMed Links

Gene Details

Gene
CPPED1
Gene Name
calcineurin like phosphoesterase domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001099455.1 537 Silent Mutation ACC,ACT T142T NP_001092925.1
NM_018340.2 537 Silent Mutation ACC,ACT T284T NP_060810.2

View Full Product Details