Product Details

SNP ID
rs192563150
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:55531277 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCGCGCCCAGCGAGAGCCCCACAC[A/G]GAACTCGTACAGGAAGCCACGGAGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SBK2 PubMed Links

Gene Details

Gene
SBK2
Gene Name
SH3 domain binding kinase family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001101401.2 337 Missense Mutation CGT,TGT R108C NP_001094871.2
XM_006723327.3 337 Missense Mutation CGT,TGT R108C XP_006723390.1
XM_011527227.2 337 Missense Mutation CGT,TGT R108C XP_011525529.1
Gene
SBK3
Gene Name
SH3 domain binding kinase family member 3
There are no transcripts associated with this gene.

View Full Product Details