Product Details

SNP ID
rs192673888
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.17:81899528 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCACAGATCAAGTGTCTGCAGATGG[A/G]CCCAGGGCCTGTAGGTCAGACATTG
Phenotype
MIM: 604171 MIM: 614534 MIM: 607996 MIM: 602679
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
ALYREF PubMed Links

Gene Details

Gene
ALYREF
Gene Name
Aly/REF export factor
There are no transcripts associated with this gene.

Gene
ANAPC11
Gene Name
anaphase promoting complex subunit 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001002244.2 558 Silent Mutation GGA,GGG G132G NP_001002244.1
NM_001002245.2 558 Intron NP_001002245.1
NM_001002246.2 558 Intron NP_001002246.1
NM_001002247.2 558 Intron NP_001002247.1
NM_001002248.2 558 Intron NP_001002248.1
NM_001002249.2 558 Intron NP_001002249.1
NM_001289414.1 558 Intron NP_001276343.1
NM_001289415.1 558 Intron NP_001276344.1
NM_001289416.1 558 Intron NP_001276345.1
NM_001289417.1 558 Intron NP_001276346.1
NM_001289418.1 558 Intron NP_001276347.1
NM_001289419.1 558 Intron NP_001276348.1
NM_001289420.1 558 Intron NP_001276349.1
NM_016476.11 558 Intron NP_057560.8
Gene
NPB
Gene Name
neuropeptide B
There are no transcripts associated with this gene.

Gene
PCYT2
Gene Name
phosphate cytidylyltransferase 2, ethanolamine
There are no transcripts associated with this gene.

View Full Product Details