Product Details

SNP ID
rs192696675
Assay Type
Functionally tested
NCBI dbSNP Submissions
13
Location
Chr.1:56854927 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGTTTGCTGTTGTTTTCTTCATCT[C/T]GTCTTTGATGACTTGTCAGCCTGGG
Phenotype
MIM: 120950
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C8A PubMed Links

Gene Details

Gene
C8A
Gene Name
complement component 8 alpha subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000562.2 132 Missense Mutation TCG,TTG S9L NP_000553.1
XM_011542079.2 132 Missense Mutation TCG,TTG S9L XP_011540381.1
XM_017002234.1 132 Missense Mutation TCG,TTG S9L XP_016857723.1

View Full Product Details