Product Details
- SNP ID
-
rs192692700
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:120222721 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGCAGCAGGTGGATGCCCACAGCGA[C/T]CAGCATGGCTTTGAACTCCACACAG
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TMEM185B
PubMed Links
Gene Details
- Gene
- TMEM185B
- Gene Name
- transmembrane protein 185B
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_024121.2 |
688 |
Missense Mutation |
ATC,GTC |
I86V |
NP_077026.2 |
View Full Product Details