Product Details

SNP ID
rs192593829
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:102319876 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGTCGCACGTGTGGCCCCGCTGGG[A/C]CCCGAGCAGCTGCGGCAGGTCCTGG
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ZNF839 PubMed Links

Gene Details

Gene
ZNF839
Gene Name
zinc finger protein 839
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001267827.1 126 Intron NP_001254756.1
NM_001267828.1 126 Intron NP_001254757.1
NM_018335.4 126 Silent Mutation GGA,GGC G37G NP_060805.3
XM_006720203.3 126 Intron XP_006720266.2
XM_011536945.1 126 Silent Mutation GGA,GGC G37G XP_011535247.1
XM_011536946.2 126 Silent Mutation GGA,GGC G37G XP_011535248.1
XM_011536948.2 126 Intron XP_011535250.1
XM_011536949.2 126 Silent Mutation GGA,GGC G37G XP_011535251.1
XM_011536950.2 126 Silent Mutation GGA,GGC G37G XP_011535252.1
XM_017021447.1 126 Silent Mutation GGA,GGC G37G XP_016876936.1
XM_017021448.1 126 Silent Mutation GGA,GGC G37G XP_016876937.1
XM_017021449.1 126 Intron XP_016876938.1
XM_017021450.1 126 Intron XP_016876939.1
XM_017021451.1 126 Intron XP_016876940.1

View Full Product Details