Product Details

SNP ID
rs192585482
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:104986457 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGCAAGGCCGGCGGGAGCTGGGGG[G/T]AGAGCCTTTGAGTGACCTCCAGGAG
Phenotype
MIM: 608570
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
AHNAK2 PubMed Links

Gene Details

Gene
AHNAK2
Gene Name
AHNAK nucleoprotein 2
There are no transcripts associated with this gene.

Gene
C14orf79
Gene Name
chromosome 14 open reading frame 79
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_174891.3 179 Missense Mutation GGA,GTA G9V NP_777551.2
XM_005267317.3 179 Missense Mutation GGA,GTA G9V XP_005267374.1
XM_005267318.3 179 Missense Mutation GGA,GTA G9V XP_005267375.1

View Full Product Details