Product Details
- SNP ID
-
rs192640536
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:68874876 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGGGTGGATTAGAGGTCACTCTCGC[C/T]GTACAGCGCCGTGGAGAAGGACATG
- Phenotype
-
MIM: 102575
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ACTN1
PubMed Links
Gene Details
- Gene
- ACTN1
- Gene Name
- actinin alpha 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001102.3 |
3614 |
Missense Mutation |
AGC,GGC |
S888G |
NP_001093.1 |
NM_001130004.1 |
3614 |
Missense Mutation |
AGC,GGC |
S910G |
NP_001123476.1 |
NM_001130005.1 |
3614 |
Missense Mutation |
AGC,GGC |
S883G |
NP_001123477.1 |
XM_011537265.2 |
3614 |
Missense Mutation |
AGC,GGC |
S925G |
XP_011535567.1 |
XM_011537266.2 |
3614 |
Missense Mutation |
AGC,GGC |
S920G |
XP_011535568.1 |
XM_011537267.2 |
3614 |
Missense Mutation |
AGC,GGC |
S917G |
XP_011535569.1 |
XM_011537268.2 |
3614 |
Missense Mutation |
AGC,GGC |
S912G |
XP_011535570.1 |
XM_017021720.1 |
3614 |
Missense Mutation |
AGC,GGC |
S1102G |
XP_016877209.1 |
XM_017021721.1 |
3614 |
Missense Mutation |
AGC,GGC |
S1094G |
XP_016877210.1 |
XM_017021722.1 |
3614 |
Missense Mutation |
AGC,GGC |
S1073G |
XP_016877211.1 |
XM_017021723.1 |
3614 |
Missense Mutation |
AGC,GGC |
S1065G |
XP_016877212.1 |
XM_017021725.1 |
3614 |
Missense Mutation |
AGC,GGC |
S1052G |
XP_016877214.1 |
XM_017021726.1 |
3614 |
Missense Mutation |
AGC,GGC |
S1044G |
XP_016877215.1 |
XM_017021727.1 |
3614 |
Missense Mutation |
AGC,GGC |
S891G |
XP_016877216.1 |
XM_017021728.1 |
3614 |
Missense Mutation |
AGC,GGC |
S862G |
XP_016877217.1 |
View Full Product Details