Product Details

SNP ID
rs192580494
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:2641827 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCTGCCACTGGCCAGGACAGACAG[C/T]GAACAGTAGCCCAGCGCCTGCCTGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MMEL1 PubMed Links

Gene Details

Gene
MMEL1
Gene Name
membrane metallo-endopeptidase-like 1
There are no transcripts associated with this gene.

Gene
TTC34
Gene Name
tetratricopeptide repeat domain 34
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242672.1 1316 Silent Mutation TCA,TCG S414S NP_001229601.1
XM_016999990.1 1316 Silent Mutation TCA,TCG S414S XP_016855479.1

View Full Product Details