Product Details

SNP ID
rs192437065
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:49156217 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAATAGGCATATTTTGGGAAACTTT[C/T]CACTTTCTGTAGTTGGTGAATTTGC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C10orf128 PubMed Links

Gene Details

Gene
C10orf128
Gene Name
chromosome 10 open reading frame 128
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010863.2 557 Intron NP_001010863.1
NM_001288740.1 557 Intron NP_001275669.1
NM_001288741.1 557 Intron NP_001275670.1
NM_001288742.1 557 Missense Mutation GAA,GGA E102G NP_001275671.1
NM_001288743.1 557 Missense Mutation AAA,GAA K165E NP_001275672.1
XM_011539378.2 557 Intron XP_011537680.1
XM_011539379.1 557 Intron XP_011537681.1
XM_011539380.1 557 Intron XP_011537682.1
XM_011539382.1 557 Intron XP_011537684.1
XM_011539384.1 557 Intron XP_011537686.1
XM_011539385.1 557 Intron XP_011537687.1
XM_017015792.1 557 Intron XP_016871281.1
XM_017015793.1 557 Intron XP_016871282.1
XM_017015794.1 557 Intron XP_016871283.1
XM_017015795.1 557 Intron XP_016871284.1
XM_017015796.1 557 Intron XP_016871285.1
XM_017015797.1 557 Intron XP_016871286.1
Gene
FAM170B-AS1
Gene Name
FAM170B antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details