Product Details

SNP ID
rs192499967
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:66975507 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGCAAGAGAAGTCCAGGTCCCAG[A/G]GCCTCGAGAGAGGCACAGAAGGGCC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C11orf86 PubMed Links

Gene Details

Gene
C11orf86
Gene Name
chromosome 11 open reading frame 86
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136485.1 225 Missense Mutation AGC,GGC S49G NP_001129957.1

View Full Product Details