Product Details

SNP ID
rs192454033
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:99816956 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTAGAAGGGAAGATGAGCACCCAG[A/G]ATCCCTCAGATCTGTGGAGCAGATC
Phenotype
MIM: 605768
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DAPP1 PubMed Links

Gene Details

Gene
DAPP1
Gene Name
dual adaptor of phosphotyrosine and 3-phosphoinositides 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001306151.1 127 Missense Mutation AAT,GAT N15D NP_001293080.1
NM_014395.2 127 Missense Mutation AAT,GAT N15D NP_055210.2
XM_011531840.2 127 Missense Mutation AAT,GAT N15D XP_011530142.1
XM_011531842.1 127 Missense Mutation AAT,GAT N15D XP_011530144.1
XM_011531843.2 127 Intron XP_011530145.1
XM_017008023.1 127 Missense Mutation AAT,GAT N15D XP_016863512.1
XM_017008024.1 127 Missense Mutation AAT,GAT N15D XP_016863513.1

View Full Product Details