Product Details

SNP ID
rs192235966
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:58709952 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCACAAGGCCATGGAGCCGGTATTC[C/T]GGCAAGGTGCCTGCCATTCTCATCT
Phenotype
MIM: 607424
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GLYAT PubMed Links

Gene Details

Gene
GLYAT
Gene Name
glycine-N-acyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005838.3 923 Intron NP_005829.3
NM_201648.2 923 Silent Mutation CCA,CCG P235P NP_964011.2
XM_017017087.1 923 Silent Mutation CCA,CCG P171P XP_016872576.1

View Full Product Details