Product Details

SNP ID
rs192172039
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:36750420 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGTTTAACAAGATAGGAAAAGTGA[C/T]GAAAGGCCTTCCCACACTCCTTACA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF850 PubMed Links

Gene Details

Gene
ZNF850
Gene Name
zinc finger protein 850
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193552.1 683 Missense Mutation CAT,CGT H207R NP_001180481.1
NM_001267779.1 683 Missense Mutation CAT,CGT H175R NP_001254708.1

View Full Product Details