Product Details

SNP ID
rs192850755
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:35744127 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGAGTTCAGCCACGGCCCGCTCTC[C/T]ATCCTCCTCCCTCCGGAACTGCAGG
Phenotype
MIM: 614143 MIM: 606834 MIM: 607632 MIM: 601080
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IGFLR1 PubMed Links

Gene Details

Gene
IGFLR1
Gene Name
IGF like family receptor 1
There are no transcripts associated with this gene.

Gene
KMT2B
Gene Name
lysine methyltransferase 2B
There are no transcripts associated with this gene.

Gene
LIN37
Gene Name
lin-37 DREAM MuvB core complex component
There are no transcripts associated with this gene.

Gene
PSENEN
Gene Name
presenilin enhancer gamma-secretase subunit
There are no transcripts associated with this gene.

Gene
U2AF1L4
Gene Name
U2 small nuclear RNA auxiliary factor 1 like 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040425.2 301 Missense Mutation AGA,GGA R84G NP_001035515.1
NM_144987.3 301 Missense Mutation AGA,GGA R84G NP_659424.2

View Full Product Details