Product Details

SNP ID
rs192844575
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:19219384 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCAAAAACCAGGCCAAAGAGGAC[A/G]CTGCTGATTAACACGTAGTCATAGT
Phenotype
MIM: 616846 MIM: 609890
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EMC1 PubMed Links

Gene Details

Gene
EMC1
Gene Name
ER membrane protein complex subunit 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271427.1 2951 Silent Mutation AGC,AGT S966S NP_001258356.1
NM_001271428.1 2951 Silent Mutation AGC,AGT S966S NP_001258357.1
NM_001271429.1 2951 Silent Mutation AGC,AGT S945S NP_001258358.1
NM_015047.2 2951 Silent Mutation AGC,AGT S967S NP_055862.1
XM_005245787.1 2951 Silent Mutation AGC,AGT S970S XP_005245844.1
XM_005245788.1 2951 Silent Mutation AGC,AGT S969S XP_005245845.1
Gene
LOC101927895
Gene Name
uncharacterized LOC101927895
There are no transcripts associated with this gene.

Gene
UBR4
Gene Name
ubiquitin protein ligase E3 component n-recognin 4
There are no transcripts associated with this gene.

View Full Product Details