Product Details

SNP ID
rs193093066
Assay Type
Functionally Tested
NCBI dbSNP Submissions
6
Location
Chr.1:224114234 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTCTACCCAGCGACAGCCTCCACC[A/G]CCCGCGCCACAGCACCCGCAGCCGG
Phenotype
MIM: 609100
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
FBXO28 PubMed Links

Gene Details

Gene
FBXO28
Gene Name
F-box protein 28
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136115.2 148 Silent Mutation CCA,CCG P35P NP_001129587.1
NM_015176.3 148 Silent Mutation CCA,CCG P35P NP_055991.1

View Full Product Details