Product Details
- SNP ID
-
rs193099422
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:42104960 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGAACTCTGGCGGCGGTGGGATGAC[C/T]TCGAAGTTGAACTCCTCCTCCTCCT
- Phenotype
-
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
C6orf132
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs150599390] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- C6orf132
- Gene Name
- chromosome 6 open reading frame 132
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001164446.2 |
3485 |
Silent Mutation |
GAA,GAG |
E984E |
NP_001157918.1 |
View Full Product Details