Product Details

SNP ID
rs193240312
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:36819767 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCGGGGCAGCGGTGGGGGCAGCA[C/T]GAGCAAGACCTGCACCTACGAAGGC
Phenotype
MIM: 601861
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RFXAP PubMed Links

Gene Details

Gene
RFXAP
Gene Name
regulatory factor X associated protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000538.3 566 Missense Mutation ACG,ATG T137M NP_000529.1

View Full Product Details