Product Details
- SNP ID
-
rs193135919
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:99698564 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTGGTCAGAAGACAGGTCTGAGTCC[C/T]GACAATCCATTTCTGGATCCCTGCC
- Phenotype
-
MIM: 603646
MIM: 616753
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
COX15
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs369470330] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- COX15
- Gene Name
- COX15, cytochrome c oxidase assembly homolog
- Gene
- ENTPD7
- Gene Name
- ectonucleoside triphosphate diphosphohydrolase 7
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_020354.3 |
1219 |
Silent Mutation |
CCC,CCT |
P347P |
NP_065087.1 |
View Full Product Details