Product Details

SNP ID
rs192852831
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:129461122 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCAGAGCTGCAGTTCGCCCCCGAC[C/G]GCGAGGAGTGGGAAGTCGTGTTTCC
Phenotype
MIM: 607513
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ADAMTS19 PubMed Links

Gene Details

Gene
ADAMTS19
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_011543246.2 Intron XP_011541548.1
XM_011543247.2 Intron XP_011541549.1
XM_011543248.2 Intron XP_011541550.1
XM_011543249.2 Intron XP_011541551.1
XM_017009174.1 Intron XP_016864663.1
XM_017009175.1 Intron XP_016864664.1
XM_017009176.1 Intron XP_016864665.1
Gene
ADAMTS19-AS1
Gene Name
ADAMTS19 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details