Product Details

SNP ID
rs192915815
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:50371729 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAATTCCCAAGATTACGCACCTCTG[A/G]AACAAAGATTGGATTCATCCAGTCA
Phenotype
MIM: 300837
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DGKK PubMed Links

Gene Details

Gene
DGKK
Gene Name
diacylglycerol kinase kappa
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001013742.3 3692 Missense Mutation CCA,TCA P1203S NP_001013764.1
XM_017029268.1 3692 Missense Mutation CCA,TCA P1174S XP_016884757.1

View Full Product Details