Product Details

SNP ID
rs192885582
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:730900 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGGGGCAGCGCCCTCGTTTGAGCC[G/T]CTGCACCAGGTTCTGGATGTTGCGG
Phenotype
MIM: 614666 MIM: 611118
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CCDC78 PubMed Links

Gene Details

Gene
CCDC78
Gene Name
coiled-coil domain containing 78
There are no transcripts associated with this gene.

Gene
FAM173A
Gene Name
family with sequence similarity 173 member A
There are no transcripts associated with this gene.

Gene
HAGHL
Gene Name
hydroxyacylglutathione hydrolase-like
There are no transcripts associated with this gene.

Gene
NARFL
Gene Name
nuclear prelamin A recognition factor like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304799.1 1188 Silent Mutation AGG,CGG R277R NP_001291728.1
NM_022493.2 1188 Silent Mutation AGG,CGG R379R NP_071938.1

View Full Product Details