Product Details

SNP ID
rs192945884
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:130097206 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGCTGGTGAAACATGAAGAAAAC[C/T]GCCTGGTCTTTGAGGACTCAGAGGA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ALG1L2 PubMed Links

Gene Details

Gene
ALG1L2
Gene Name
ALG1, chitobiosyldiphosphodolichol beta-mannosyltransferase like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136152.1 Intron NP_001129624.1
Gene
FAM86HP
Gene Name
family with sequence similarity 86, member A pseudogene
There are no transcripts associated with this gene.

View Full Product Details