Product Details

SNP ID
rs192929781
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:149604002 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTTACATACAAGCCAGGGCCAGTG[A/G]ATGATCTTGTCCAGTCTTAAGGCAA
Phenotype
MIM: 300031
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HSFX2 PubMed Links

Gene Details

Gene
HSFX2
Gene Name
heat shock transcription factor family, X-linked 2
There are no transcripts associated with this gene.

Gene
TMEM185A
Gene Name
transmembrane protein 185A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001174092.2 647 Silent Mutation ATC,ATT I105I NP_001167563.1
NM_001282302.1 647 Silent Mutation ATC,ATT I164I NP_001269231.1
NM_032508.3 647 Silent Mutation ATC,ATT I164I NP_115897.1

View Full Product Details