Product Details

SNP ID
rs192959143
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:73425134 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATCAATGTAGGAATCTTATAGTCA[C/T]GAAGTAGCAGCAGGGTGGGCAGCCA
Phenotype
MIM: 614773
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MSS51 PubMed Links

Gene Details

Gene
MSS51
Gene Name
MSS51 mitochondrial translational activator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001024593.1 1193 Missense Mutation NP_001019764.1

View Full Product Details