Product Details

SNP ID
rs193042691
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:75579393 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACTTACATCCCTCTGCTCCTGCCA[A/G]TGGCTTGGCTGCTGGAGAAGTTCGG
Phenotype
MIM: 610865
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FLVCR2 PubMed Links

Gene Details

Gene
FLVCR2
Gene Name
feline leukemia virus subgroup C cellular receptor family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195283.1 797 Intron NP_001182212.1
NM_017791.2 797 Missense Mutation ATG,GTG M141V NP_060261.2
Gene
LOC102724153
Gene Name
uncharacterized LOC102724153
There are no transcripts associated with this gene.

View Full Product Details