Product Details

SNP ID
rs193035596
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:73813712 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCACAACTATGTTGTCTGGGGTTAC[C/T]GGCCAGAGCCATATCTGTACTTCAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C2orf78 PubMed Links

Gene Details

Gene
C2orf78
Gene Name
chromosome 2 open reading frame 78
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080474.1 454 Silent Mutation ACC,ACT T111T NP_001073943.1
XM_005264302.3 454 Silent Mutation ACC,ACT T111T XP_005264359.1

View Full Product Details