Product Details

SNP ID
rs193056478
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:20006480 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTTTGCCACATTCTTCACATTTA[C/T]AGGGTTTCTCTCCCGTATGGGTTCT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF682 PubMed Links

Gene Details

Gene
ZNF682
Gene Name
zinc finger protein 682
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001077349.1 1249 Intron NP_001070817.1
NM_033196.2 1249 Intron NP_149973.1
XM_017027455.1 1249 Missense Mutation TAT,TGT Y347C XP_016882944.1
XM_017027456.1 1249 Intron XP_016882945.1

View Full Product Details