Product Details

SNP ID
rs192743323
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:112162768 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTTTGTCTGCCCAGCAAATTGTTT[C/T]TTTTTTTTTTTTTTTAACCATTTTT
Phenotype
MIM: 613197
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HECTD4 PubMed Links

Gene Details

Gene
HECTD4
Gene Name
HECT domain E3 ubiquitin protein ligase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001109662.3 Intron NP_001103132.3
Gene
MIR6861
Gene Name
microRNA 6861
There are no transcripts associated with this gene.

Gene
TRAFD1
Gene Name
TRAF-type zinc finger domain containing 1
There are no transcripts associated with this gene.

View Full Product Details