Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001136505.1 | 2278 | Silent Mutation | CCG,CCT | P722P | NP_001129977.1 |
XM_011523004.2 | 2278 | Silent Mutation | XP_011521306.1 | ||
XM_011523005.2 | 2278 | Silent Mutation | XP_011521307.1 | ||
XM_011523006.2 | 2278 | Silent Mutation | XP_011521308.1 | ||
XM_011523007.2 | 2278 | Silent Mutation | XP_011521309.1 | ||
XM_011523008.2 | 2278 | Silent Mutation | XP_011521310.1 | ||
XM_011523009.2 | 2278 | Silent Mutation | XP_011521311.1 | ||
XM_011523010.2 | 2278 | Silent Mutation | XP_011521312.1 | ||
XM_011523011.2 | 2278 | Silent Mutation | XP_011521313.1 | ||
XM_011523012.2 | 2278 | Silent Mutation | XP_011521314.1 | ||
XM_011523014.2 | 2278 | Silent Mutation | XP_011521316.1 | ||
XM_011523015.2 | 2278 | Intron | XP_011521317.1 | ||
XM_011523016.2 | 2278 | Intron | XP_011521318.1 | ||
XM_011523017.1 | 2278 | Intron | XP_011521319.1 | ||
XM_011523018.1 | 2278 | Intron | XP_011521320.1 | ||
XM_011523020.1 | 2278 | Intron | XP_011521322.1 | ||
XM_017023157.1 | 2278 | Intron | XP_016878646.1 | ||
XM_017023158.1 | 2278 | Intron | XP_016878647.1 | ||
XM_017023159.1 | 2278 | Intron | XP_016878648.1 |