Product Details

SNP ID
rs192728116
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:58155837 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCATCCTGGTGATGCTGAGGTCCCA[C/T]TCGGGAAAGGCAAGGAGGAAGGCAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MSX2P1 PubMed Links

Gene Details

Gene
MSX2P1
Gene Name
msh homeobox 2 pseudogene 1
There are no transcripts associated with this gene.

Gene
OR4D1
Gene Name
olfactory receptor family 4 subfamily D member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012374.1 684 Silent Mutation CAC,CAT H228H NP_036506.1

View Full Product Details