Product Details

SNP ID
rs192725409
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:20162504 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGAATTGTATATATTAGATAACA[C/T]GAGAAAACATGGAAAGATATGACAA
Phenotype
MIM: 300075
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RPS6KA3 PubMed Links

Gene Details

Gene
RPS6KA3
Gene Name
ribosomal protein S6 kinase A3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004586.2 Intron NP_004577.1
XM_005274573.2 Intron XP_005274630.1
XM_005274577.2 Intron XP_005274634.1
XM_006724507.3 Intron XP_006724570.1
XM_011545555.1 Intron XP_011543857.1
XM_011545556.1 Intron XP_011543858.1
XM_011545557.2 Intron XP_011543859.1
XM_011545558.2 Intron XP_011543860.1
XM_011545559.1 Intron XP_011543861.1
XM_011545560.1 Intron XP_011543862.1
XM_011545561.1 Intron XP_011543863.1
XM_011545562.1 Intron XP_011543864.1
XM_011545563.2 Intron XP_011543865.1
XM_017029713.1 Intron XP_016885202.1
XM_017029714.1 Intron XP_016885203.1
XM_017029715.1 Intron XP_016885204.1
XM_017029716.1 Intron XP_016885205.1
XM_017029717.1 Intron XP_016885206.1
XM_017029718.1 Intron XP_016885207.1
XM_017029719.1 Intron XP_016885208.1

View Full Product Details