Product Details

SNP ID
rs192783899
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:114790809 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGATCAGCAGGAATTTGTTGGTATA[A/G]TTCATTCGGTTCTTGGTGAAGGCCA
Phenotype
MIM: 604052
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TNFSF15 PubMed Links

Gene Details

Gene
TNFSF15
Gene Name
tumor necrosis factor superfamily member 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204344.1 515 Silent Mutation AAC,AAT N74N NP_001191273.1
NM_005118.3 515 Silent Mutation AAC,AAT N133N NP_005109.2

View Full Product Details