Product Details

SNP ID
rs217816
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:37294294 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATCTACAAAGTTACTTTAATTAAA[G/T]AAAATAATTTTATGTTATTTAATAT
Phenotype
MIM: 606694
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
NUP155 PubMed Links

Gene Details

Gene
NUP155
Gene Name
nucleoporin 155
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278312.1 Intron NP_001265241.1
NM_004298.3 Intron NP_004289.1
NM_153485.2 Intron NP_705618.1
XM_011514165.2 Intron XP_011512467.1

View Full Product Details